Assay Details
| Gene Symbol: Ttc30b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001127607.1 | NP_001121079.1 | 1 | 1219 | 71 | |
| GenBank mRNA | BC166739.1 | - | 1 | 1219 | 7937387 | 71 |
More Information
Set Membership: |
|

Ttc30b
Genomic Map