Assay Details
| Gene Symbol: Kcna4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_012971.2 | NP_037103.1 | 1 | 1820 | 69 | |
| XM_006234673.2 | XP_006234735.1 | 2 | 3016 | 69 | ||
| XM_006234674.2 | XP_006234736.1 | 2 | 2868 | 69 | ||
| GenBank mRNA | M32867.1 | - | 1 | 1820 | 69 | |
| X16002.1 | - | 1 | 2234 | 69 | ||
More Information
Set Membership: |
|

Kcna4
Genomic Map