Assay Details
| Gene Symbol: Timd2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001013855.1 | NP_001013877.1 | 2 - 3 | 98 | 85 | |
| XM_006246168.1 | XP_006246230.1 | 3 - 4 | 383 | 85 | ||
| XM_008767667.1 | XP_008765889.1 | 3 - 4 | 146 | 85 | ||
| XM_008767668.1 | XP_008765890.1 | 3 - 4 | 146 | 85 | ||
| XM_008767669.1 | XP_008765891.1 | 3 - 4 | 638 | 85 | ||
| GenBank mRNA | AY255103.1 | - | 2 - 3 | 475 | 85 | |
| BC089832.1 | - | 2 - 3 | 98 | 7324518 | 85 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the AY255103.1, BC089832.1, NM_001013855.1, XM_006246168.1, XM_008767669.1 transcripts.
Set Membership: |
|

Timd2
Genomic Map