Assay Details
| Gene Symbol: Uimc1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001013884.1 | NP_001013906.1 | 4 - 5 | 427 | 59 | |
| XM_006253613.2 | XP_006253675.1 | 4 - 5 | 831 | 59 | ||
| XM_006253616.2 | XP_006253678.1 | 4 - 5 | 830 | 59 | ||
| GenBank mRNA | BC087149.1 | - | 4 - 5 | 427 | 7131093 | 59 |
More Information
Set Membership: |
|

Uimc1
Genomic Map