Assay Details
| Gene Symbol: Uso1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_019379.1 | NP_062252.1 | 12 - 13 | 1337 | 57 | |
| XM_006250750.2 | XP_006250812.1 | 12 - 13 | 1358 | 57 | ||
| GenBank mRNA | FQ209746.1 | - | 4 - 5 | 526 | 57 | |
| U14192.1 | - | 12 - 13 | 1252 | 57 | ||
| U15589.1 | - | 12 - 13 | 1337 | 57 | ||
More Information
Set Membership: |
|

Uso1
Genomic Map