Assay Details
| Gene Symbol: Hspa2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_021863.3 | NP_068635.1 | 1 | 522 | 77 | |
| XM_006240247.2 | XP_006240309.1 | 2 | 499 | 77 | ||
| GenBank mRNA | BC081803.1 | - | 2 | 493 | 7112083 | 77 |
More Information
Set Membership: |
|

Hspa2
Genomic Map