Assay Details
| Gene Symbol: Pbld1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_138530.2 | NP_612539.2 | 693 | 62 | ||
| XM_006256372.2 | XP_006256434.1 | 8 - 9 | 1008 | 62 | ||
| GenBank mRNA | BC078735.1 | - | 693 | 7126368 | 62 | |
| FQ209640.1 | - | 728 | 62 | |||
More Information
Set Membership: |
|

Pbld1
Genomic Map