Assay Details
| Gene Symbol: Slc16a2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_147216.1 | NP_671749.1 | 5 - 6 | 1416 | 83 | |
| GenBank mRNA | AJ496570.1 | - | 5 - 6 | 1416 | 83 | |
More Information
Set Membership: |
|

Slc16a2
Genomic Map