Assay Details
| Gene Symbol: Osm | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001006961.1 | NP_001006962.1 | 1 - 2 | 35 | 62 | |
| GenBank mRNA | AB167521.1 | - | 1 - 2 | 35 | 62 | |
More Information
Set Membership: |
|

Osm
Genomic Map