Assay Details
| Gene Symbol: Lhfpl4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_181387.2 | NP_852052.1 | 3 - 4 | 823 | 65 | |
| GenBank mRNA | AY278321.1 | - | 3 - 4 | 823 | 65 | |
More Information
Set Membership: |
|

Lhfpl4
Genomic Map