Assay Details
| Gene Symbol: Slfnl1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001024347.1 | NP_001019518.1 | 2 - 3 | 794 | 60 | |
| XM_008764060.1 | XP_008762282.1 | 1 - 2 | 1103 | 60 | ||
| XM_008764061.1 | XP_008762283.1 | 3 - 4 | 2328 | 60 | ||
| XM_008764062.1 | XP_008762284.1 | 3 - 4 | 2328 | 60 | ||
| XM_008764063.1 | XP_008762285.1 | 1 - 2 | 1103 | 60 | ||
| GenBank mRNA | BC079280.1 | - | 2 - 3 | 794 | 7134187 | 60 |
More Information
Set Membership: |
|

Slfnl1
Genomic Map