Assay Details
| Gene Symbol: St8sia5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_213628.1 | NP_998793.1 | 3 - 4 | 310 | 73 | |
| XM_006254968.2 | XP_006255030.1 | 4 - 5 | 696 | 73 | ||
| GenBank mRNA | AJ699422.1 | - | 3 - 4 | 310 | 73 | |
More Information
Set Membership: |
|

St8sia5
Genomic Map