Assay Details
Gene Symbol: Calu | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001033898.1 | NP_001029070.1 | 6 - 7 | 894 | 87 | |
NM_022535.2 | NP_071980.2 | 6 - 7 | 894 | 87 | ||
GenBank mRNA | AJ001929.1 | - | 6 - 7 | 927 | 87 | |
BC101908.1 | - | 6 - 7 | 894 | 7933832 | 87 | |
FQ222101.1 | - | 5 - 6 | 737 | 87 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
