Assay Details
| Gene Symbol: Asb12 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001037367.1 | NP_001032444.1 | 2 - 3 | 219 | 107 | |
| GenBank mRNA | BC107911.1 | - | 2 - 3 | 219 | 7453889 | 107 |
More Information
Additional Information:
This assay is in the 5'UTR region of the BC107911.1, NM_001037367.1 transcripts.
Set Membership: |
|

Asb12
Genomic Map