Assay Details
| Gene Symbol: Rbm43 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001037649.1 | NP_001032738.1 | 4 - 5 | 432 | 88 | |
| XM_006234167.2 | XP_006234229.1 | 4 - 5 | 496 | 88 | ||
| XM_006234168.2 | XP_006234230.1 | 4 - 5 | 1294 | 88 | ||
| XM_008761743.1 | XP_008759965.1 | 4 - 5 | 1298 | 88 | ||
| XM_008761744.1 | XP_008759966.1 | 4 - 5 | 500 | 88 | ||
| GenBank mRNA | BC107648.1 | - | 4 - 5 | 432 | 7930358 | 88 |
More Information
Set Membership: |
|

Rbm43
Genomic Map