Assay Details
| Gene Symbol: Prelid1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001009636.1 | NP_001009636.1 | 1 - 2 | 225 | 77 | |
| GenBank mRNA | BC088284.1 | - | 1 - 2 | 225 | 7323653 | 77 |
| FQ212767.1 | - | 1 - 2 | 237 | 77 | ||
| FQ226295.1 | - | 1 - 2 | 239 | 77 | ||
More Information
Set Membership: |
|

Prelid1
Genomic Map