Assay Details
| Gene Symbol: Nmb | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001109149.1 | NP_001102619.1 | 1 - 2 | 219 | 78 | |
| XM_006229445.2 | XP_006229507.1 | 1 - 2 | 893 | 78 | ||
| XM_008759554.1 | XP_008757776.1 | 1 - 2 | 893 | 78 | ||
More Information
Set Membership: |
|

Nmb
Genomic Map