Assay Details
| Gene Symbol: Bbs10 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001109286.1 | NP_001102756.1 | 1 - 2 | 228 | 125 | |
| GenBank mRNA | BC160915.1 | - | 1 - 2 | 220 | 7457522 | 125 |
| FQ211675.1 | - | 1 - 2 | 210 | 125 | ||
More Information
Set Membership: |
|

Bbs10
Genomic Map