Assay Details
Gene Symbol: Rpl19 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_031103.1 | NP_112365.1 | 2 - 3 | 141 | 106 | |
GenBank mRNA | BC058135.1 | - | 2 - 3 | 141 | 6918377 | 106 |
FQ222282.1 | - | 2 - 3 | 106 | |||
J02650.1 | - | 2 - 3 | 141 | 106 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
