Assay Details
| Gene Symbol: Tmem252 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001007738.1 | NP_001007739.1 | 2 | 520 | 62 | |
| GenBank mRNA | BC079347.1 | - | 2 | 520 | 7107289 | 62 |
More Information
Set Membership: |
|

Tmem252
Genomic Map