Assay Details
| Gene Symbol: Ccdc8 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001009533.2 | NP_001009533.2 | 1 | 3227 | 109 | |
| GenBank mRNA | BC060520.1 | - | 2 | 3203 | 5597839 | 109 |
More Information
Set Membership: |
|

Ccdc8
Genomic Map