Assay Details
| Gene Symbol: Hs3st3a1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001172067.1 | NP_001165538.1 | 1 | 119 | 111 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

Hs3st3a1| Gene Symbol: Hs3st3a1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001172067.1 | NP_001165538.1 | 1 | 119 | 111 | |
Set Membership: |
|