Assay Details
Gene Symbol: LOC688542 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_008760597.1 | XP_008758819.1 | 1012 | 80 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
