Assay Details
Gene Symbol: GAL2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001181968.1 | NP_013182.1 | 1 | 1651 | 82 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
