Assay Details
| Gene Symbol: FBXO28 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003130584.6 | XP_003130632.2 | 572 | 87 | ||
| GenBank mRNA | AK343237.1 | - | 3 - 4 | 539 | 87 | |
More Information
Set Membership: |
|

FBXO28
Genomic Map