Assay Details
| Gene Symbol: PCGF6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_021073477.1 | XP_020929136.1 | 691 | 119 | ||
| XM_021073478.1 | XP_020929137.1 | 690 | 119 | |||
| XM_021073479.1 | XP_020929138.1 | 690 | 119 | |||
| XM_021073480.1 | XP_020929139.1 | 690 | 119 | |||
More Information
Set Membership: |
|

PCGF6
Genomic Map