Assay Details
| Gene Symbol: SAR1A | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001031786.1 | NP_001026956.1 | 5 - 6 | 475 | 65 | |
| XM_005657393.3 | XP_005657450.1 | 637 | 65 | |||
| XM_005657394.3 | XP_005657451.1 | 665 | 65 | |||
| GenBank mRNA | AK234361.1 | - | 6 - 7 | 600 | 65 | |
| AK350329.1 | - | 6 - 7 | 590 | 65 | ||
| AK399576.1 | - | 6 - 7 | 603 | 65 | ||
| AY996812.1 | - | 5 - 6 | 475 | 65 | ||
More Information
Set Membership: |
|

SAR1A
Genomic Map