Assay Details
Gene Symbol: EFNB2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001114286.1 | NP_001107758.1 | 2 - 3 | 408 | 104 | |
GenBank mRNA | EF682141.1 | - | 2 - 3 | 408 | 104 | |
KF500033.1 | - | 2 - 3 | 496 | 104 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
