Assay Details
| Gene Symbol: VPS52 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_001925393.5 | XP_001925428.2 | 954 | 74 | ||
| XM_013977772.2 | XP_013833226.1 | 393 | 74 | |||
| XM_021098592.1 | XP_020954251.1 | 296 | 74 | |||
| GenBank mRNA | AK232658.1 | - | 2 - 3 | 1242 | 74 | |
| AK239403.1 | - | 3 - 4 | 416 | 74 | ||
| AK397809.1 | - | 3 - 4 | 479 | 74 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the XM_013977772.2, XM_021098592.1 transcripts.
Set Membership: |
|

VPS52
Genomic Map