Assay Details
| Gene Symbol: PRELID1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001244208.1 | NP_001231137.1 | 2 - 3 | 521 | 64 | |
| GenBank mRNA | AK238807.1 | - | 2 - 3 | 519 | 64 | |
| AK351591.1 | - | 2 - 3 | 486 | 64 | ||
| AK351936.1 | - | 2 - 3 | 479 | 64 | ||
| AK392733.1 | - | 2 - 3 | 494 | 64 | ||
| AK396232.1 | - | 2 - 3 | 479 | 64 | ||
| AK397156.1 | - | 2 - 3 | 484 | 64 | ||
| AK397840.1 | - | 2 - 3 | 521 | 64 | ||
| AK398185.1 | - | 2 - 3 | 479 | 64 | ||
| CJ022605.1 | - | 2 - 3 | 521 | 64 | ||
More Information
Set Membership: |
|

PRELID1
Genomic Map