Assay Details
| Gene Symbol: MAT1A | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001243187.1 | NP_001230116.1 | 2 - 3 | 309 | 59 | |
| GenBank mRNA | AK346080.1 | - | 2 - 3 | 309 | 59 | |
More Information
Set Membership: |
|

MAT1A
Genomic Map