Assay Details
| Gene Symbol: CHMP3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003354734.4 | XP_003354782.3 | 479 | 86 | ||
| XM_013995984.2 | XP_013851438.1 | 563 | 86 | |||
| GenBank mRNA | AK231203.1 | - | 3 - 4 | 362 | 86 | |
| AK232282.1 | - | 3 - 4 | 393 | 86 | ||
| AK400025.1 | - | 3 - 4 | 481 | 86 | ||
| AY609625.1 | - | 3 - 4 | 547 | 86 | ||
More Information
Set Membership: |
|

CHMP3
Genomic Map