Assay Details
Gene Symbol: BAK1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_001928147.4 | XP_001928182.1 | 496 | 70 | ||
XM_013977773.2 | XP_013833227.1 | 771 | 70 | |||
XM_021098603.1 | XP_020954262.1 | 619 | 70 | |||
XM_021098604.1 | XP_020954263.1 | 355 | 70 | |||
GenBank mRNA | AJ001204.1 | - | 1 - 2 | 58 | 70 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
