Assay Details
| Gene Symbol: NODAL | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_021073885.1 | XP_020929544.1 | 1686 | 57 | ||
| GenBank mRNA | AM072821.1 | - | 1 | 557 | 57 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.
| Gene Symbol: NODAL | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_021073885.1 | XP_020929544.1 | 1686 | 57 | ||
| GenBank mRNA | AM072821.1 | - | 1 | 557 | 57 | |
Set Membership: |
|