Assay Details
| Gene Symbol: RAB5C | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001244254.1 | NP_001231183.1 | 6 | 928 | 57 | |
| GenBank mRNA | AK235644.1 | - | 6 | 895 | 57 | |
| AK393049.1 | - | 6 | 902 | 57 | ||
| AK400630.1 | - | 6 | 918 | 57 | ||
| AY550060.1 | - | 1 | 151 | 57 | ||
| BX922371.2 | - | 3 | 393 | 57 | ||
| EW384073.2 | - | 2 | 204 | 57 | ||
More Information
Set Membership: |
|

RAB5C
Genomic Map