Assay Details
Gene Symbol: GCM1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_001927486.7 | XP_001927521.4 | 589 | 74 | ||
XM_021098829.1 | XP_020954488.1 | 566 | 74 | |||
XM_021098830.1 | XP_020954489.1 | 512 | 74 | |||
GenBank mRNA | AK345878.1 | - | 1 - 2 | 546 | 74 | |
AK349097.1 | - | 3 - 4 | 569 | 74 | ||
AY594287.1 | - | 1 - 2 | 113 | 74 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
