Assay Details
| Gene Symbol: TNFSF11 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_001925694.6 | XP_001925729.4 | 585 | 100 | ||
| GenBank mRNA | DQ523616.1 | - | 2 - 3 | 226 | 100 | |
More Information
Set Membership: |
|

TNFSF11
Genomic Map