Assay Details
| Gene Symbol: CYP26A1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001315792.1 | NP_001302721.1 | 5 - 6 | 1057 | 74 | |
| XM_021071775.1 | XP_020927434.1 | 3098 | 74 | |||
| GenBank mRNA | EF625351.1 | - | 4 - 5 | 609 | 74 | |
| KP687250.1 | - | 5 - 6 | 1057 | 74 | ||
More Information
Set Membership: |
|

CYP26A1
Genomic Map