Assay Details
| Gene Symbol: AHSG | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005652369.3 | XP_005652426.1 | 1288 | 60 | ||
| GenBank mRNA | AK233033.1 | - | 6 - 7 | 844 | 60 | |
| X56021.1 | - | 758 | 60 | |||
More Information
Set Membership: |
|

AHSG
Genomic Map