Assay Details
| Gene Symbol: HTR2B | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001164019.1 | NP_001157491.1 | 2 | 605 | 71 | |
| XM_005672269.3 | XP_005672326.1 | 878 | 71 | |||
| XM_013984563.2 | XP_013840017.1 | 402 | 71 | |||
| GenBank mRNA | Z48174.1 | - | 45 | 71 | ||
More Information
Set Membership: |
|

HTR2B
Genomic Map