Assay Details
| Gene Symbol: ART5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005653658.3 | XP_005653715.1 | 1131 | 63 | ||
| XM_005653659.3 | XP_005653716.1 | 1139 | 63 | |||
| XM_021062457.1 | XP_020918116.1 | 1016 | 63 | |||
| XM_021062458.1 | XP_020918117.1 | 801 | 63 | |||
| GenBank mRNA | AJ294468.1 | - | 1 | 454 | 63 | |
More Information
Set Membership: |
|

ART5
Genomic Map