Assay Details
| Gene Symbol: CYP4F2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK233364.1 | - | 5 | 546 | 74 | |
| AK394199.1 | - | 5 | 588 | 74 | ||
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.
| Gene Symbol: CYP4F2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK233364.1 | - | 5 | 546 | 74 | |
| AK394199.1 | - | 5 | 588 | 74 | ||
Set Membership: |
|