Assay Details
| Gene Symbol: CYP4F2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003123433.3 | XP_003123481.1 | 1643 | 115 | ||
| XM_003123434.4 | XP_003123482.1 | 1643 | 115 | |||
| GenBank mRNA | AK233483.1 | - | 13 | 1555 | 115 | |
More Information
Set Membership: |
|

CYP4F2
Genomic Map