Assay Details
| Gene Symbol: CYP4F2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK233364.1 | - | 5 | 508 | 96 | |
| AK394199.1 | - | 5 | 550 | 96 | ||
| EF625356.1 | - | 4 | 344 | 96 | ||
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.
| Gene Symbol: CYP4F2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK233364.1 | - | 5 | 508 | 96 | |
| AK394199.1 | - | 5 | 550 | 96 | ||
| EF625356.1 | - | 4 | 344 | 96 | ||
Set Membership: |
|