Assay Details
| Gene Symbol: GDI2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001001643.1 | NP_001001643.1 | 10 | 1353 | 79 | |
| GenBank mRNA | AK343512.1 | - | 10 | 1411 | 79 | |
| AK349700.1 | - | 6 | 636 | 79 | ||
| AK399966.1 | - | 10 | 1384 | 79 | ||
| AY553929.1 | - | 10 | 1353 | 79 | ||
More Information
Set Membership: |
|

GDI2
Genomic Map