Assay Details
| Gene Symbol: LMCD1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001008692.1 | NP_001008692.1 | 6 | 1050 | 78 | |
| XM_021068617.1 | XP_020924276.1 | 1035 | 78 | |||
| GenBank mRNA | AK236730.1 | - | 6 | 1113 | 78 | |
| AK395882.1 | - | 6 | 1117 | 78 | ||
| AY610047.1 | - | 6 | 1072 | 78 | ||
| AY821789.1 | - | 6 | 1050 | 78 | ||
More Information
Set Membership: |
|

LMCD1
Genomic Map