Assay Details
| Gene Symbol: TMEM59 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001044558.1 | NP_001038023.1 | 8 | 952 | 97 | |
| XM_005665403.3 | XP_005665460.1 | 959 | 97 | |||
| XM_005665404.3 | XP_005665461.1 | 986 | 97 | |||
| XM_005665405.3 | XP_005665462.1 | 1012 | 97 | |||
| GenBank mRNA | AK234544.1 | - | 8 | 835 | 97 | |
| AK234840.1 | - | 8 | 889 | 97 | ||
| AK397575.1 | - | 9 | 882 | 97 | ||
| DQ200860.1 | - | 8 | 952 | 97 | ||
More Information
Set Membership: |
|

TMEM59
Genomic Map