Assay Details
Gene Symbol: KISS1R | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001044624.2 | NP_001038089.1 | 6 | 1049 | 68 | |
GenBank mRNA | DQ459346.1 | - | 6 | 1050 | 68 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() |
Panther Classification:
