Assay Details
| Gene Symbol: JPT1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001048069.1 | NP_001041534.1 | 3 | 246 | 101 | |
| XM_013980752.2 | XP_013836206.1 | 365 | 116 | |||
| GenBank mRNA | AK233714.1 | - | 3 | 331 | 101 | |
| AY610380.1 | - | 3 | 286 | 101 | ||
More Information
Set Membership: |
|

JPT1
Genomic Map