Assay Details
| Gene Symbol: DCN | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_213920.1 | NP_999085.1 | 8 | 1251 | 67 | |
| GenBank mRNA | AF125537.1 | - | 8 | 1251 | 67 | |
| AF140270.1 | - | 8 | 1137 | 67 | ||
| AK232120.1 | - | 8 | 1251 | 67 | ||
| AK232219.1 | - | 8 | 1156 | 67 | ||
| AK394646.1 | - | 5 | 639 | 67 | ||
| AK395892.1 | - | 8 | 1229 | 67 | ||
| AK395963.1 | - | 8 | 1440 | 67 | ||
| AK397441.1 | - | 8 | 1435 | 67 | ||
| AK399316.1 | - | 8 | 1248 | 67 | ||
More Information
Set Membership: |
|

DCN
Genomic Map